siRNA Details
Gene Symbol: Fam126b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025710.1 | NP_001020881.1 | 7, 8 | 641 | ||
XM_006244983.2 | XP_006245045.1 | 6, 7 | 648 | |||
XM_006244984.2 | XP_006245046.1 | 6, 7 | 647 | |||
XM_006244985.2 | XP_006245047.1 | 7, 8 | 681 | |||
XM_008767100.1 | XP_008765322.1 | 6, 7 | 1285 | |||
XM_008767101.1 | XP_008765323.1 | 7, 8 | 897 | |||
XM_008767102.1 | XP_008765324.1 | 7, 8 | 682 | |||
XM_008767103.1 | XP_008765325.1 | 6, 7 | 1196 | |||
XM_008767104.1 | XP_008765326.1 | 6, 7 | 645 | |||
XM_008767105.1 | XP_008765327.1 | 6, 7 | 648 | |||
GenBank mRNA | BC097998.1 | - | 7, 8 | 641 | 7453086 |
