siRNA Details
Gene Symbol: Tmbim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007713.1 | NP_001007714.1 | 5 | 491 | ||
XM_008767238.1 | XP_008765460.1 | 5 | 2863 | |||
XM_008767239.1 | XP_008765461.1 | 6 | 514 | |||
GenBank mRNA | BC079087.1 | - | 5 | 491 | 7123233 |
