siRNA Details
Gene Symbol: Tmbim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007713.1 | NP_001007714.1 | 7, 8 | 594 | ||
XM_008767238.1 | XP_008765460.1 | 7, 8 | 2966 | |||
XM_008767239.1 | XP_008765461.1 | 8, 9 | 617 | |||
GenBank mRNA | BC079087.1 | - | 7, 8 | 594 | 7123233 |
