siRNA Details
Gene Symbol: NEWGENE_1308624 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245535.2 | XP_006245597.1 | 2 | 1776 | ||
XM_008767394.1 | XP_008765616.1 | 3 | 397 |
Gene Symbol: Neu4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108234.1 | NP_001101704.1 | 416 | |||
XM_008767376.1 | XP_008765598.1 | 1776 |
