siRNA Details
Gene Symbol: Mettl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191814.1 | NP_001178743.1 | 1 | 125 | ||
XM_006245658.2 | XP_006245720.1 | 2 | 783 | |||
XM_006245661.2 | XP_006245723.1 | 2 | 784 |
