siRNA Details
Gene Symbol: Magee1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079891.1 | NP_001073360.1 | 1 | 1203 | ||
GenBank mRNA | BC128754.1 | - | 1 | 1203 | 7103930 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Magee1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079891.1 | NP_001073360.1 | 1 | 1203 | ||
GenBank mRNA | BC128754.1 | - | 1 | 1203 | 7103930 |