siRNA Details
Gene Symbol: Bcor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191586.1 | NP_001178515.1 | 4 | 2854 | ||
XM_006256659.2 | XP_006256721.1 | 4 | 2814 | |||
XM_006256660.2 | XP_006256722.1 | 4 | 2884 | |||
XM_006256661.2 | XP_006256723.1 | 4 | 2978 | |||
XM_006256663.2 | XP_006256725.1 | 4 | 2922 | |||
XM_006256664.2 | XP_006256726.1 | 4 | 2922 | |||
XM_006256665.2 | XP_006256727.1 | 4 | 2922 | |||
XM_008773074.1 | XP_008771296.1 | 4 | 8422 |
