siRNA Details
Gene Symbol: Smarca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271190.1 | NP_001258119.1 | 6 | 925 | ||
XM_006257525.2 | XP_006257587.2 | 7 | 927 | |||
XM_006257526.2 | XP_006257588.2 | 7 | 942 | |||
XM_006257527.2 | XP_006257589.2 | 7 | 942 | |||
XM_008773524.1 | XP_008771746.1 | 7 | 942 | |||
XM_008773526.1 | XP_008771748.1 | 7 | 895 |
