siRNA Details
Gene Symbol: Htatsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108259.1 | NP_001101729.1 | 2 | 223 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Htatsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108259.1 | NP_001101729.1 | 2 | 223 |