siRNA Details
Gene Symbol: Kif15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181635.2 | NP_853666.1 | 17 | 2094 | ||
GenBank mRNA | AY291580.1 | - | 17 | 2094 | ||
AY291581.1 | - | 17 | 2094 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kif15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181635.2 | NP_853666.1 | 17 | 2094 | ||
GenBank mRNA | AY291580.1 | - | 17 | 2094 | ||
AY291581.1 | - | 17 | 2094 |