siRNA Details
Gene Symbol: Dfna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191749.1 | NP_001178678.1 | 7 | 1047 | ||
GenBank mRNA | AY194291.1 | - | 2 | 167 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dfna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191749.1 | NP_001178678.1 | 7 | 1047 | ||
GenBank mRNA | AY194291.1 | - | 2 | 167 |