siRNA Details
Gene Symbol: Usp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024790.1 | NP_001019961.1 | 13 | 1333 | ||
XM_006245756.2 | XP_006245818.1 | 14 | 1545 | |||
XM_006245757.2 | XP_006245819.1 | 13 | 2382 | |||
XM_006245758.2 | XP_006245820.1 | 13 | 1475 | |||
XM_006245759.1 | XP_006245821.1 | 13 | 1426 | |||
GenBank mRNA | AY641530.1 | - | 13 | 1333 |
