siRNA Details
Gene Symbol: Usp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024790.1 | NP_001019961.1 | 13, 14 | 1431 | ||
XM_006245756.2 | XP_006245818.1 | 14, 15 | 1643 | |||
XM_006245757.2 | XP_006245819.1 | 13, 14 | 2480 | |||
XM_006245758.2 | XP_006245820.1 | 13, 14 | 1573 | |||
XM_006245759.1 | XP_006245821.1 | 13, 14 | 1524 | |||
GenBank mRNA | AY641530.1 | - | 13, 14 | 1431 |
