siRNA Details
Gene Symbol: Linc00514 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767626.1 | XP_008765848.1 | 32 | 1198 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Linc00514 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767626.1 | XP_008765848.1 | 32 | 1198 |