siRNA Details
Gene Symbol: Traf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127548.1 | NP_001121020.1 | 10 | 923 | ||
XM_006245982.2 | XP_006246044.1 | 10 | 1017 | |||
XM_006245983.2 | XP_006246045.1 | 10 | 1012 | |||
XM_006245984.2 | XP_006246046.1 | 9 | 858 | |||
XM_006245985.2 | XP_006246047.1 | 9 | 835 | |||
XM_006245986.2 | XP_006246048.1 | 9 | 855 | |||
XM_006245987.2 | XP_006246049.1 | 9 | 832 | |||
XM_006245988.2 | XP_006246050.1 | 10 | 1054 | |||
XM_006245989.2 | XP_006246051.1 | 10 | 1051 | |||
XM_006245990.2 | XP_006246052.1 | 9 | 804 | |||
XM_006245991.2 | XP_006246053.1 | 9 | 808 | |||
XM_006245992.2 | XP_006246054.1 | 9 | 801 | |||
XM_006245993.2 | XP_006246055.1 | 8 | 794 | |||
XM_008767584.1 | XP_008765806.1 | 10 | 1070 | |||
XM_008767585.1 | XP_008765807.1 | 10 | 1067 | |||
XM_008767586.1 | XP_008765808.1 | 10 | 1065 | |||
GenBank mRNA | BC161916.1 | - | 10 | 923 | 8369808 |
