siRNA Details
Gene Symbol: Traf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127548.1 | NP_001121020.1 | 17 | 1621 | ||
XM_006245982.2 | XP_006246044.1 | 17 | 1715 | |||
XM_006245983.2 | XP_006246045.1 | 17 | 1710 | |||
XM_006245984.2 | XP_006246046.1 | 16 | 1556 | |||
XM_006245985.2 | XP_006246047.1 | 16 | 1533 | |||
XM_006245986.2 | XP_006246048.1 | 16 | 1553 | |||
XM_006245987.2 | XP_006246049.1 | 16 | 1530 | |||
XM_006245988.2 | XP_006246050.1 | 17 | 1752 | |||
XM_006245989.2 | XP_006246051.1 | 17 | 1749 | |||
XM_006245990.2 | XP_006246052.1 | 16 | 1502 | |||
XM_006245991.2 | XP_006246053.1 | 16 | 1506 | |||
XM_006245992.2 | XP_006246054.1 | 16 | 1499 | |||
XM_006245993.2 | XP_006246055.1 | 15 | 1492 | |||
XM_008767584.1 | XP_008765806.1 | 17 | 1768 | |||
XM_008767585.1 | XP_008765807.1 | 17 | 1765 | |||
XM_008767586.1 | XP_008765808.1 | 17 | 1763 | |||
GenBank mRNA | BC161916.1 | - | 17 | 1621 | 8369808 |
