siRNA Details
Gene Symbol: Traf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127548.1 | NP_001121020.1 | 19 | 1941 | ||
XM_006245982.2 | XP_006246044.1 | 19 | 2035 | |||
XM_006245983.2 | XP_006246045.1 | 19 | 2030 | |||
XM_006245984.2 | XP_006246046.1 | 18 | 1876 | |||
XM_006245985.2 | XP_006246047.1 | 18 | 1853 | |||
XM_006245986.2 | XP_006246048.1 | 18 | 1873 | |||
XM_006245987.2 | XP_006246049.1 | 18 | 1850 | |||
XM_006245988.2 | XP_006246050.1 | 19 | 2072 | |||
XM_006245989.2 | XP_006246051.1 | 19 | 2069 | |||
XM_006245990.2 | XP_006246052.1 | 18 | 1822 | |||
XM_006245991.2 | XP_006246053.1 | 18 | 1826 | |||
XM_006245992.2 | XP_006246054.1 | 18 | 1819 | |||
XM_006245993.2 | XP_006246055.1 | 17 | 1812 | |||
XM_008767584.1 | XP_008765806.1 | 19 | 2088 | |||
XM_008767585.1 | XP_008765807.1 | 19 | 2085 | |||
XM_008767586.1 | XP_008765808.1 | 19 | 2083 | |||
GenBank mRNA | BC161916.1 | - | 19 | 1941 | 8369808 |
