siRNA Details
Gene Symbol: Wdr24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191084.1 | NP_001178013.1 | 3 | 1511 | ||
XM_008767587.1 | XP_008765809.1 | 3 | 2817 |

More Information
Other Species Targeted

