siRNA Details
Gene Symbol: Wdr24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191084.1 | NP_001178013.1 | 4 | 1574 | ||
XM_008767587.1 | XP_008765809.1 | 4 | 2880 | |||
GenBank mRNA | BC098800.1 | - | 1 | 70 |

More Information
Other Species Targeted

