siRNA Details
Gene Symbol: Slc47a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014118.2 | NP_001014140.1 | 2 | 152 | ||
GenBank mRNA | AB248823.1 | - | 2 | 152 | ||
AB248824.1 | - | 2 | 152 | |||
BC088413.1 | - | 172 | 7109897 | |||
FQ217802.1 | - | 166 |
