siRNA Details
Gene Symbol: RGD1304728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767904.1 | XP_008766126.1 | 18, 19 | 2925 | ||
XM_008767905.1 | XP_008766127.1 | 18, 19 | 2841 | |||
XM_008767906.1 | XP_008766128.1 | 18, 19 | 3049 | |||
XM_008767907.1 | XP_008766129.1 | 17, 18 | 2980 | |||
XM_340837.6 | XP_340838.3 | 18, 19 | 3076 |

More Information
Other Species Targeted

