siRNA Details
Gene Symbol: Luc7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108291.1 | NP_001101761.1 | 3, 4 | 381 | ||
XM_006247155.2 | XP_006247217.1 | 3, 4 | 373 | |||
XM_006247156.2 | XP_006247218.1 | 3, 4 | 372 | |||
XM_008768078.1 | XP_008766300.1 | 3, 4 | 372 | |||
GenBank mRNA | FQ212347.1 | - | 3, 4 | 354 | ||
FQ231407.1 | - | 1, 2 | 31 | |||
FQ231682.1 | - | 3, 4 | 354 | |||
FQ235074.1 | - | 3, 4 | 347 |
