siRNA Details
Gene Symbol: Luc7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108291.1 | NP_001101761.1 | 10 | 1381 | ||
XM_006247155.2 | XP_006247217.1 | 10 | 1373 | |||
XM_006247156.2 | XP_006247218.1 | 10 | 1372 | |||
XM_008768078.1 | XP_008766300.1 | 10 | 1372 | |||
GenBank mRNA | FQ212347.1 | - | 10 | 1352 | ||
FQ235260.1 | - | 3 | 395 |
