siRNA Details
Gene Symbol: Cdc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108298.1 | NP_001101768.1 | 10, 11 | 1646 | ||
XM_006247354.2 | XP_006247416.1 | 10, 11 | 1648 | |||
XM_006247355.2 | XP_006247417.1 | 10, 11 | 1646 | |||
GenBank mRNA | FQ233913.1 | - | 1, 2 | 147 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011799, NM_001025779, AJ223087, BC052434, AK166003, and others.
Gene Ontology Categories:
