siRNA Details
Gene Symbol: Cdc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108298.1 | NP_001101768.1 | 9, 10 | 1429 | ||
XM_006247354.2 | XP_006247416.1 | 9, 10 | 1431 | |||
XM_006247355.2 | XP_006247417.1 | 9, 10 | 1429 |
