siRNA Details
Gene Symbol: Etv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108299.1 | NP_001101769.1 | 6 | 630 | ||
XM_006247369.2 | XP_006247431.1 | 6 | 703 | |||
XM_006247370.2 | XP_006247432.1 | 6 | 651 | |||
XM_006247371.2 | XP_006247433.1 | 6 | 611 | |||
XM_006247372.2 | XP_006247434.1 | 6 | 458 | |||
XM_008768085.1 | XP_008766307.1 | 6 | 700 | |||
XM_008768086.1 | XP_008766308.1 | 6 | 704 | |||
GenBank mRNA | BC168950.1 | - | 6 | 504 | 9022277 |
