siRNA Details
Gene Symbol: Etv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108299.1 | NP_001101769.1 | 8 | 848 | ||
XM_006247369.2 | XP_006247431.1 | 8 | 921 | |||
XM_006247370.2 | XP_006247432.1 | 8 | 869 | |||
XM_006247371.2 | XP_006247433.1 | 8 | 829 | |||
XM_006247372.2 | XP_006247434.1 | 8 | 676 | |||
XM_008768085.1 | XP_008766307.1 | 8 | 918 | |||
XM_008768086.1 | XP_008766308.1 | 8 | 904 | |||
GenBank mRNA | BC168950.1 | - | 8 | 722 | 9022277 |
