siRNA Details
Gene Symbol: Cdc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024793.1 | NP_001019964.1 | 7 | 895 | ||
XM_006247533.2 | XP_006247595.1 | 6 | 2613 | |||
XM_006247534.2 | XP_006247596.1 | 6 | 2613 | |||
XM_006247535.2 | XP_006247597.1 | 7 | 1029 | |||
XM_006247536.2 | XP_006247598.1 | 6 | 2614 | |||
XM_006247537.1 | XP_006247599.1 | 4 | 561 | |||
XM_008768295.1 | XP_008766517.1 | 6 | 882 | |||
GenBank mRNA | BC097475.1 | - | 7 | 895 | 7457819 |
