siRNA Details
Gene Symbol: Cd300le | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202463.1 | NP_001189392.1 | 2 | 89 | ||
GenBank mRNA | HQ263411.1 | - | 2 | 89 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd300le | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202463.1 | NP_001189392.1 | 2 | 89 | ||
GenBank mRNA | HQ263411.1 | - | 2 | 89 |