siRNA Details
Gene Symbol: Wdr45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039587.1 | NP_001034676.1 | 4 | 306 | ||
XM_006247900.2 | XP_006247962.1 | 4 | 449 | |||
GenBank mRNA | AM182327.1 | - | 4 | 306 |

More Information
Other Species Targeted

Gene Ontology Categories:
