siRNA Details
Gene Symbol: Dscr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108316.2 | NP_001101786.2 | 1 | 35 | ||
XM_006248072.2 | XP_006248134.1 | 1 | 489 | |||
XM_006248073.2 | XP_006248135.1 | 1 | 488 | |||
XM_008768583.1 | XP_008766805.1 | 1 | 489 | |||
GenBank mRNA | BC160826.1 | - | 1 | 35 | 7444837 |
