siRNA Details
Gene Symbol: Gramd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191826.1 | NP_001178755.1 | 1428 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gramd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191826.1 | NP_001178755.1 | 1428 |