siRNA Details
Gene Symbol: Tango2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108323.1 | NP_001101793.1 | 8 | 777 | ||
XM_006248628.2 | XP_006248690.1 | 8 | 902 | |||
XM_006248629.2 | XP_006248691.1 | 8 | 772 | |||
XM_006248630.2 | XP_006248692.1 | 8 | 797 | |||
XM_006248632.2 | XP_006248694.1 | 8 | 902 | |||
XM_006248633.2 | XP_006248695.1 | 8 | 901 | |||
XM_008768888.1 | XP_008767110.1 | 8 | 775 | |||
GenBank mRNA | FQ213583.1 | - | 7 | 804 |
