siRNA Details
Gene Symbol: Lztr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108326.2 | NP_001101796.1 | 20 | 2414 | ||
XM_006248707.2 | XP_006248769.1 | 20 | 2727 | |||
GenBank mRNA | BC169071.1 | - | 20 | 2414 | 7938530 | |
FQ211175.1 | - | 3 | 305 | |||
FQ211947.1 | - | 19 | 3127 | |||
FQ218020.1 | - | 5 | 533 |
