siRNA Details
Gene Symbol: Lrch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127551.1 | NP_001121023.1 | 10 | 1174 | ||
XM_006249175.2 | XP_006249237.1 | 10 | 1190 | |||
GenBank mRNA | BC166781.1 | - | 10 | 1174 | 7934984 |
