siRNA Details
Gene Symbol: Rsrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014128.1 | NP_001014150.1 | 12 | 1438 | ||
XM_006249338.2 | XP_006249400.1 | 10 | 1305 | |||
XM_006249339.2 | XP_006249401.1 | 10 | 1302 | |||
XM_006249340.2 | XP_006249402.1 | 11 | 1396 | |||
XM_006249341.2 | XP_006249403.1 | 11 | 1393 | |||
GenBank mRNA | BC087067.1 | - | 12 | 1438 | 7133034 |
