siRNA Details
Gene Symbol: Srgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134958.1 | NP_001128430.1 | 2 | 703 | ||
XM_006249782.2 | XP_006249844.1 | 2 | 790 | |||
XM_006249783.2 | XP_006249845.1 | 2 | 674 | |||
XM_006249784.2 | XP_006249846.1 | 2 | 791 | |||
XM_008769479.1 | XP_008767701.1 | 2 | 785 | |||
GenBank mRNA | BC168690.1 | - | 2 | 703 | 7934193 |
