siRNA Details
Gene Symbol: Srgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134958.1 | NP_001128430.1 | 10 | 1970 | ||
XM_006249782.2 | XP_006249844.1 | 10 | 2057 | |||
XM_006249783.2 | XP_006249845.1 | 10 | 1941 | |||
XM_006249784.2 | XP_006249846.1 | 10 | 2055 | |||
XM_006249785.2 | XP_006249847.1 | 9 | 3006 | |||
XM_008769479.1 | XP_008767701.1 | 10 | 2052 | |||
XM_008769480.1 | XP_008767702.1 | 7 | 974 | |||
XM_008769481.1 | XP_008767703.1 | 7 | 903 | |||
GenBank mRNA | BC168690.1 | - | 10 | 1970 | 7934193 | |
FQ212420.1 | - | 3 | 374 |
