siRNA Details
Gene Symbol: Uchl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012149.1 | NP_001012149.1 | 6 | 631 | ||
XM_006249960.2 | XP_006250022.1 | 6 | 666 | |||
XM_006249961.2 | XP_006250023.1 | 6 | 666 | |||
XM_006249962.2 | XP_006250024.1 | 6 | 667 | |||
XM_006249963.2 | XP_006250025.1 | 6 | 668 | |||
GenBank mRNA | BC088841.1 | - | 6 | 631 | 7125859 |
