siRNA Details
Gene Symbol: RGD1309106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014130.1 | NP_001014152.1 | 3 | 243 | ||
XM_006250168.2 | XP_006250230.1 | 3 | 232 | |||
XM_006250169.2 | XP_006250231.1 | 3 | 232 | |||
XM_006250170.2 | XP_006250232.1 | 3 | 446 | |||
XM_006250171.2 | XP_006250233.1 | 3 | 193 | |||
GenBank mRNA | BC083573.1 | - | 3 | 243 | 7112776 |
