siRNA Details
Gene Symbol: Rcsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108349.1 | NP_001101819.1 | 5, 6 | 1276 | ||
XM_006250181.2 | XP_006250243.1 | 6, 7 | 1402 | |||
XM_008769663.1 | XP_008767885.1 | 5, 6 | 1225 |
