siRNA Details
Gene Symbol: Ahctf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271270.1 | NP_001258199.1 | 22 | 3057 | ||
XM_006250386.2 | XP_006250448.1 | 22 | 3104 | |||
XM_008769858.1 | XP_008768080.1 | 22 | 2760 |
