siRNA Details
Gene Symbol: Esrrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203336.2 | NP_976081.1 | 7 | 1345 | ||
XM_006250443.2 | XP_006250505.1 | 6 | 1998 | |||
GenBank mRNA | AB126962.1 | - | 6 | 1229 | ||
AB126963.1 | - | 7 | 1457 | |||
AY341057.1 | - | 6 | 1108 |
