siRNA Details
Gene Symbol: Mtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100898.1 | NP_001094368.1 | 15 | 1698 | ||
XM_006250569.2 | XP_006250631.1 | 16 | 1893 | |||
XM_008769939.1 | XP_008768161.1 | 15 | 1603 | |||
GenBank mRNA | BC160928.1 | - | 13 | 1267 |
