siRNA Details
Gene Symbol: Mtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100898.1 | NP_001094368.1 | 7, 8 | 951 | ||
XM_006250569.2 | XP_006250631.1 | 8, 9 | 1146 | |||
XM_008769939.1 | XP_008768161.1 | 7, 8 | 856 | |||
GenBank mRNA | BC093403.1 | - | 7, 8 | 911 | ||
BC160928.1 | - | 5, 6 | 520 |
