siRNA Details
Gene Symbol: Mtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100898.1 | NP_001094368.1 | 6 | 807 | ||
XM_006250569.2 | XP_006250631.1 | 7 | 1002 | |||
XM_008769939.1 | XP_008768161.1 | 6 | 712 | |||
GenBank mRNA | BC093403.1 | - | 6 | 767 | ||
BC160928.1 | - | 4 | 376 | |||
FQ227583.1 | - | 7 | 1004 |
