siRNA Details
Gene Symbol: Cdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108352.1 | NP_001101822.1 | 11 | 1479 | ||
XM_006250570.2 | XP_006250632.1 | 9 | 1215 | |||
XM_008769940.1 | XP_008768162.1 | 9 | 1192 |
