siRNA Details
Gene Symbol: Tbrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012154.1 | NP_001012154.1 | 5 | 1182 | ||
XM_006251342.2 | XP_006251404.1 | 7 | 1364 | |||
XM_006251343.2 | XP_006251405.1 | 7 | 1294 | |||
GenBank mRNA | BC087073.1 | - | 5 | 1182 | 7134456 | |
FQ217643.1 | - | 7 | 1327 | |||
FQ217828.1 | - | 7 | 1327 |
