siRNA Details
Gene Symbol: Tbrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012154.1 | NP_001012154.1 | 1 | 195 | ||
XM_006251342.2 | XP_006251404.1 | 3 | 377 | |||
XM_006251343.2 | XP_006251405.1 | 3 | 307 | |||
GenBank mRNA | BC087073.1 | - | 1 | 195 | 7134456 | |
FQ217643.1 | - | 3 | 340 | |||
FQ217828.1 | - | 3 | 340 |
