siRNA Details
Gene Symbol: Tbrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012154.1 | NP_001012154.1 | 2 | 629 | ||
XM_006251342.2 | XP_006251404.1 | 4 | 811 | |||
XM_006251343.2 | XP_006251405.1 | 4 | 741 | |||
GenBank mRNA | BC087073.1 | - | 2 | 629 | 7134456 | |
FQ217643.1 | - | 4 | 774 | |||
FQ217828.1 | - | 4 | 774 |
